α-Cyclopropylbenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: alpha-Cyclopropylbenzyl alcohol, Cyclopropyl(phenyl)methanol, α-Cyclopropylbenzyl alcohol, Cyclopropyl phenyl carbinol, Cyclopropylphenylcarbinol

235048-5G 235048-25G
77490-210EA 60.77 USD
77490-210 77500-402
α-Cyclopropylbenzyl alcohol ≥95%
α-Cyclopropylbenzyl alcohol

Precaución: For research use only

Formula: C₁₀H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00001299
CAS Number: 1007-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR