2-Acetyl-6-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(6-Methoxy-2-pyridinyl)ethanone

220098-5G 220098-25G
77490-002EA 83.43 USD
77490-002 77500-252
2-Acetyl-6-methoxypyridine ≥95%
2-Acetyl-6-methoxypyridine

Precaución: For research use only

Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276836
CAS Number: 21190-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR