3-Bromo-5-phenoxypyridine ≥97%

Supplier: AOBChem USA

Warning

21428-5G 21428-25G
77489-914EA 117.42 USD
77489-914 77500-174
3-Bromo-5-phenoxypyridine ≥97%
3-Bromo-5-phenoxypyridine

Precaución: For research use only

Formula: C₁₁H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD10698710
CAS Number: 28232-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR