N-(3-Formyl-pyridin-2-yl)-2,2-dimethylpropionamide ≥97%

Supplier: AOBChem USA

Warning

20549-5G 20549-25G
77489-694EA 163.77 USD
77489-694 77500-000
N-(3-Formyl-pyridin-2-yl)-2,2-dimethylpropionamide ≥97%
N-(3-Formyl-pyridin-2-yl)-2,2-dimethylpropionamide

Precaución: For research use only

Formula: C₁₁H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01830170
CAS Number: 86847-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR