2,5-Dichloro-3-formylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dichloronicotinaldehyde, 2,5-Dichloro-3-formylpyridine

20186-5G 20186-25G
77489-602EA 150.38 USD
77489-602 77499-924
2,5-Dichloro-3-formylpyridine ≥97%
2,5-Dichloro-3-formylpyridine

Precaución: For research use only

Formula: C₆H₃Cl₂NO
MW: 176.0000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056627
CAS Number: 176433-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR