3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-methox-5-(trifluoromethyl)ypyridine, 3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine

20083-5G 20083-25G
77489-588EA 90.64 USD
77489-588 77499-912
3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine ≥95%
3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine

Precaución: For research use only

Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972202
CAS Number: 124432-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR