1-Bromo-2-chloro-4-pyrrolidinobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Bromo-3-chlorophenyl)pyrrolidine

193073-5G 193073-25G
77489-486EA 173.04 USD
77489-486 77499-834
1-Bromo-2-chloro-4-pyrrolidinobenzene ≥95%
1-Bromo-2-chloro-4-pyrrolidinobenzene

Precaución: For research use only

Formula: C₁₀H₁₁BrClN
MW: 260.558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18837620
CAS Number: 1291487-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR