Pentafluoroethyl phenyl ketone ≥97%

Supplier: AOBChem USA

Warning

182375-5G 182375-25G
77489-100EA 96.82 USD
77489-100 77499-544
Pentafluoroethyl phenyl ketone ≥97%
Pentafluoroethyl phenyl ketone

Precaución: For research use only

Formula: C₉H₅F₅O
Storage Temperature: Ambient
MDL Number: MFCD00018062
CAS Number: 394-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR