1-Fluoro-4-methyl-2-propoxybenzene ≥96%

Supplier: AOBChem USA

Warning

Synonyms: Benzene, 1-fluoro-4-methyl-2-propoxy-

182325-5G 182325-25G
77489-066EA 88.58 USD
77489-066 77499-520
1-Fluoro-4-methyl-2-propoxybenzene ≥96%
1-Fluoro-4-methyl-2-propoxybenzene

Precaución: For research use only

Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19237203
CAS Number: 1311197-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR