1-Bromo-2,4-dimethyl-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

180364-5G 180364-25G
77488-984EA 125.66 USD
77488-984 77499-472
1-Bromo-2,4-dimethyl-3-nitrobenzene ≥97%
1-Bromo-2,4-dimethyl-3-nitrobenzene

Precaución: For research use only

Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD00092653
CAS Number: 39053-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR