1,4-Dibromo-2,5-bis(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1,4-Bis(trifluoromethyl)-2,5-dibromobenzene, 1,4-Dibromo-2,5-bis(trifluoromethyl)benzene

17558-5G 17558-25G
77488-894EA 61.8 USD
77488-894 77499-390
1,4-Dibromo-2,5-bis(trifluoromethyl)benzene ≥95%
1,4-Dibromo-2,5-bis(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₈H₂Br₂F₆
MW: 371.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15526888
CAS Number: 2375-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR