Methyl 2-fluoro-4-methyl-5-nitrobenzoate ≥95%

Supplier: AOBChem USA

Warning

171746-5G 171746-25G
77488-800EA 113.3 USD
77488-800 77499-304
Methyl 2-fluoro-4-methyl-5-nitrobenzoate ≥95%
Methyl 2-fluoro-4-methyl-5-nitrobenzoate

Precaución: For research use only

Formula: C₉H₈FNO₄
Storage Temperature: Ambient
MDL Number: MFCD23863743
CAS Number: 753924-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR