Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 5-bromobenzo[b]thiophene-2-carboxylate, Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate

169596-5G 169596-25G
77488-716EA 67.98 USD
77488-716 77499-234
Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate ≥95%
Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate

Precaución: For research use only

Formula: C₁₁H₉BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD12756246
CAS Number: 13771-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR