1-(2,5-Dimethylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

168680-5G 168680-25G
77488-684EA 92.7 USD
77488-684 77499-216
1-(2,5-Dimethylphenyl)ethanol ≥95%
1-(2,5-Dimethylphenyl)ethanol

Precaución: For research use only

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00016855
CAS Number: 32917-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR