3-Iodo-5-(trifluoromethyl)-2-pyridinylamine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Iodo-5-(trifluoromethyl)pyridin-2-amine, 2-Amino-3-iodo-5-(trifluoromethyl)pyridine

167107-5G 167107-25G
77488-582EA 95.79 USD
77488-582 77499-158
3-Iodo-5-(trifluoromethyl)-2-pyridinylamine ≥97%
3-Iodo-5-(trifluoromethyl)-2-pyridinylamine

Precaución: For research use only

Formula: C₆H₄F₃IN₂
MW: 288.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697686
CAS Number: 211308-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR