3-Phenyl-3-pentanol ≥97%

Supplier: AOBChem USA

Warning

164655-5G 164655-25G
77488-310EA 153.47 USD
77488-310 77498-978
3-Phenyl-3-pentanol ≥97%
3-Phenyl-3-pentanol

Precaución: For research use only

Formula: C₆H₅C(OH)(CH₂CH₃)₂
MW: 164.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021830
CAS Number: 1565-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR