3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16196-5G 16196-25G
77488-188EA 107.12 USD
77488-188 77498-882
3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde ≥97%
3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₈H₃ClF₄O
MW: 226.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631343
CAS Number: 186517-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR