4,6-Dichloro-5-fluoronicotinic acid ≥95%

Supplier: AOBChem USA

Warning

158050-5G 158050-25G
77488-088EA 84.46 USD
77488-088 77498-788
4,6-Dichloro-5-fluoronicotinic acid ≥95%
4,6-Dichloro-5-fluoronicotinic acid

Precaución: For research use only

Formula: C₆H₂Cl₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD18256593
CAS Number: 154012-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR