Methyl-3-methylbenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

155713-5G 155713-25G
77488-036EA 124.63 USD
77488-036 77498-738
Methyl-3-methylbenzo[b]thiophene-2-carboxylate ≥97%
Methyl 3-methylbenzo[b]thiophene-2-carboxylate

Precaución: For research use only

Formula: C₁₁H₁₀O₂S
Storage Temperature: Ambient
MDL Number: MFCD02683110
CAS Number: 3133-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR