tert-Butyl 4-(3-bromobenzoyl)piperazine-1-carboxylate ≥97%

Supplier: AOBChem USA

Warning

154839-5G 154839-25G
77488-020EA 73.13 USD
77488-020 77498-712
tert-Butyl 4-(3-bromobenzoyl)piperazine-1-carboxylate ≥97%
tert-Butyl 4-(3-bromobenzoyl)piperazine-1-carboxylate

Precaución: For research use only

Formula: C₁₆H₂₁BrN₂O₃
MW: 369.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368338
CAS Number: 890153-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR