2,3-Dibromo-α,α,α-trifluorotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dibromobenzotrifluoride

15165-5G 15165-25G
77487-970EA 160.68 USD
77487-970 77498-658
2,3-Dibromo-α,α,α-trifluorotoluene ≥97%
2,3-Dibromo-α,α,α-trifluorotoluene

Precaución: For research use only

Formula: C₇H₃Br₂F₃
MW: 303.902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093035
CAS Number: 493038-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR