2-Bromo-5-fluoro-4-methylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

14894-5G 14894-25G
77487-906EA 120.51 USD
77487-906 77498-606
2-Bromo-5-fluoro-4-methylbenzonitrile ≥95%
2-Bromo-5-fluoro-4-methylbenzonitrile

Precaución: For research use only

Formula: C₈H₅BrFN
MW: 214.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782072
CAS Number: 916792-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR