2,4-Dimethoxyphenol ≥96%

Supplier: AOBChem USA

Warning

142599-5G 142599-25G
77487-782EA 60.77 USD
77487-782 77498-498
2,4-Dimethoxyphenol ≥96%
2,4-Dimethoxyphenol

Precaución: For research use only

Formula: C₈H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00060637
CAS Number: 13330-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR