(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,2-Difluoro-1,3-benzodioxole-5-boronic acid, (2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid

13597-5G 13597-25G
77487-574EA 177.16 USD
77487-574 77498-356
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid ≥97%
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid

Precaución: For research use only

Formula: C₇H₅BF₂O₄
Storage Temperature: Ambient
MDL Number: MFCD08706236
CAS Number: 190903-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR