4-Fluororesorcinol ≥96%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-Fluoro-2,4-dihydroxybenzene
127998-5G
127998-25G
77487-372EA
71.07
USD
77487-372
77498-170
4-Fluororesorcinol ≥96%
4-Fluororesorcinol
Precaución: For research use only
Formula:
C₆H₅FO₂ MW: 128.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03789074 CAS Number: 103068-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...