3-(Ethylthio)phenylboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
12360-5G
12360-25G
77487-224EA
129.78
USD
77487-224
77498-048
3-(Ethylthio)phenylboronic acid ≥95%
3-(Ethylthio)phenylboronic acid
Precaución: For research use only
Formula:
C₈H₁₁BO₂S MW: 182.05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03092930 CAS Number: 870718-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...