3-Chloroquinoline ≥97%

Supplier: AOBChem USA

Warning

11487-5G 11487-25G
77486-884EA 75.19 USD
77486-884 77497-748
3-Chloroquinoline ≥97%
3-Chloroquinoline

Precaución: For research use only

Formula: C9H6ClN
MW: 163.6036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234489
CAS Number: 612-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR