4-(Cyclopentylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Cyclopentylaminocarbonyl)phenylboronic acid

11286-5G 11286-25G
77486-862EA 131.84 USD
77486-862 77497-720
4-(Cyclopentylcarbamoyl)phenylboronic acid ≥97%
4-(Cyclopentylcarbamoyl)phenylboronic acid

Precaución: For research use only

Formula: C₁₂H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD03788418
CAS Number: 850568-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR