2,4-Bis(trifluoromethyl)benzamide ≥95%

Supplier: AOBChem USA

Warning

108654-5G 108654-25G
77486-786EA 118.45 USD
77486-786 77497-628
2,4-Bis(trifluoromethyl)benzamide ≥95%
2,4-Bis(trifluoromethyl)benzamide

Precaución: For research use only

Formula: C₉H₅F₆NO
MW: 257.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083536
CAS Number: 53130-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR