5-Bromo-7-methylbenzo[b]thiophene-2-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

99039-5G 99039-25G
77497-184EA 125.66 USD
77497-184 77506-068
5-Bromo-7-methylbenzo[b]thiophene-2-carboxylic acid ≥95%
5-Bromo-7-methylbenzo[b]thiophene-2-carboxylic acid

Precaución: For research use only

Formula: C₁₀H₇BrO₂S
MW: 271.1304 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29784001
CAS Number: 1935615-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR