3-Cyano-2-fluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
95125-5G
95125-25G
77497-118EA
76.22
USD
77497-118
77506-018
3-Cyano-2-fluorobenzoic acid ≥97%
3-Cyano-2-fluorobenzoic acid
Precaución: For research use only
Formula:
C₈H₄FNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09261249 CAS Number: 219519-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...