2-Bromo-5-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

84307-5G 84307-25G
77496-708EA 86.52 USD
77496-708 77505-718
2-Bromo-5-fluoro-4-methylbenzaldehyde ≥97%
2-Bromo-5-fluoro-4-methylbenzaldehyde

Precaución: For research use only

Formula: C₈H₆BrFO
MW: 217.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782047
CAS Number: 916792-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR