Ethyl-3-fluoro-4-methylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-fluoro-4-methylphenyl)-2-oxoacetate

82428-5G 82428-25G
77496-624EA 81.37 USD
77496-624 77505-650
Ethyl-3-fluoro-4-methylbenzoylformate ≥95%
Ethyl 3-fluoro-4-methylbenzoylformate

Precaución: For research use only

Formula: C₁₁H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD01319622
CAS Number: 732251-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR