5-Bromo-1-methyl-1H-pyrazole ≥97%
Supplier: AOBChem USA
|
Warning
|
80002-5G
80002-25G
77496-506EA
75.19
USD
77496-506
77505-566
5-Bromo-1-methyl-1H-pyrazole ≥97%
5-Bromo-1-methyl-1H-pyrazole
Precaución: For research use only
Formula:
C₄H₅BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD11656572 CAS Number: 361476-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...