2,3,6-Trimethoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

77385-5G 77385-25G
77496-374EA 85.49 USD
77496-374 77505-464
2,3,6-Trimethoxybenzoic acid ≥95%
2,3,6-Trimethoxybenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00092587
CAS Number: 60241-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR