(2,4-Difluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

69577-5G 69577-25G
77496-012EA 99.91 USD
77496-012 77505-214
(2,4-Difluorophenyl)trimethylsilane ≥95%
(2,4-Difluorophenyl)trimethylsilane

Precaución: For research use only

Formula: C₉H₁₂F₂Si
Storage Temperature: Ambient
MDL Number: MFCD22682801
CAS Number: 148854-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR