4'-Bromo-2,2,2,2'-tetrafluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

65950-5G 65950-25G
77495-708EA 75.19 USD
77495-708 77504-984
4'-Bromo-2,2,2,2'-tetrafluoroacetophenone ≥95%
4'-Bromo-2,2,2,2'-tetrafluoroacetophenone

Precaución: For research use only

Formula: C₈H₃BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD19439730
CAS Number: 617706-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR