2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

63464-5G 63464-25G
77495-492EA 122.57 USD
77495-492 77504-816
2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone ≥97%
2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone

Precaución: For research use only

Formula: C₈H₃BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD08062425
CAS Number: 886502-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR