1-tert-Butyl-3-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-tert-Butyliodobenzene, 3-tert-Butyliodbenzol, 1-(tert-Butyl)-3-iodobenzene

63111-5G 63111-25G
77495-458EA 77.25 USD
77495-458 77504-796
1-tert-Butyl-3-iodobenzene ≥95%
1-tert-Butyl-3-iodobenzene

Precaución: For research use only

Formula: C₁₀H₁₃I
Storage Temperature: Ambient
MDL Number: MFCD01764504
CAS Number: 58164-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR