1-Bromo-2-fluoro-4-(methylsulfonyl)benzene ≥95%

Supplier: AOBChem USA

Warning

62602-5G 62602-25G
77495-414EA 71.07 USD
77495-414 77504-768
1-Bromo-2-fluoro-4-(methylsulfonyl)benzene ≥95%
1-Bromo-2-fluoro-4-(methylsulphonyl)benzene

Precaución: For research use only

Formula: C₇H₆BrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD11053812
CAS Number: 1032825-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR