3-Chloro-4-iodophenol ≥97%

Supplier: AOBChem USA

Warning

56788-5G 56788-25G
77495-136EA 173.04 USD
77495-136 77504-580
3-Chloro-4-iodophenol ≥97%
3-Chloro-4-iodophenol

Precaución: For research use only

Formula: C₆H₄ClIO
Storage Temperature: Ambient
MDL Number: MFCD17000018
CAS Number: 855403-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR