3-Chloro-5-nitrobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

54197-5G 54197-25G
77494-990EA 105.06 USD
77494-990 77504-460
3-Chloro-5-nitrobenzoic acid ≥95%
3-Chloro-5-nitrobenzoic acid

Precaución: For research use only

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263224
CAS Number: 34662-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR