1-Bromo-5-(difluoromethyl)-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-(difluoromethyl)-1-fluorobenzene

53467-5G 53467-25G
77494-940EA 79.31 USD
77494-940 77504-420
1-Bromo-5-(difluoromethyl)-2-fluorobenzene ≥97%
1-Bromo-5-(difluoromethyl)-2-fluorobenzene

Precaución: For research use only

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276255
CAS Number: 886509-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR