2-Fluoro-4-iodophenol ≥97%

Supplier: AOBChem USA

Warning

53041-5G 53041-25G
77494-910EA 90.64 USD
77494-910 77504-396
2-Fluoro-4-iodophenol ≥97%
2-Fluoro-4-iodophenol

Precaución: For research use only

Formula: C₆H₄FIO
Storage Temperature: Ambient
MDL Number: MFCD11109769
CAS Number: 2713-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR