3,5-Dichloro-4-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49953-5G 49953-25G
77494-690EA 129.78 USD
77494-690 77504-252
3,5-Dichloro-4-fluorobenzaldehyde ≥97%
3,5-Dichloro-4-fluorobenzaldehyde

Precaución: For research use only

Formula: C₇H₃Cl₂FO
MW: 193.0000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845864
CAS Number: 117820-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR