1-Bromo-3,5-dimethoxy-4-fluorobenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
48739-5G
48739-25G
77494-612EA
78.28
USD
77494-612
77504-198
1-Bromo-3,5-dimethoxy-4-fluorobenzene ≥95%
1-Bromo-3,5-dimethoxy-4-fluorobenzene
Precaución: For research use only
Formula:
C₈H₈BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28738436 CAS Number: 204654-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...