3,6-Dichloro-2-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48491-5G 48491-25G
77494-594EA 60.77 USD
77494-594 77504-180
3,6-Dichloro-2-hydroxybenzoic acid ≥97%
3,6-Dichloro-2-hydroxybenzoic acid

Precaución: For research use only

Formula: C₇H₄Cl₂O₃
MW: 207.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00155381
CAS Number: 3401-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR