3-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester, 3-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, tert-Butyl 4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperazine-1-carboxylate

39138-5G 39138-25G
77494-134EA 152.44 USD
77494-134 77503-836
3-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester ≥95%
3-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₂₂H₃₃BN₂O₅
Storage Temperature: Ambient
MDL Number: MFCD09266185
CAS Number: 883738-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR