3-Bromo-9-methyl-9H-carbazole ≥97%

Supplier: AOBChem USA

Warning

38047-5G 38047-25G
77494-046EA 88.58 USD
77494-046 77503-762
3-Bromo-9-methyl-9H-carbazole ≥97%
3-Bromo-9-methyl-9H-carbazole

Precaución: For research use only

Formula: C₁₃H₁₀BrN
MW: 260.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00276495
CAS Number: 91828-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR