3,4-Dibromophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

36883-5G 36883-25G
77493-760EA 182.31 USD
77493-760 77503-514
3,4-Dibromophenylboronic acid ≥97%
3,4-Dibromophenylboronic acid

Precaución: For research use only

Formula: C₆H₅BBr₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18383692
CAS Number: 1228180-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR